Assay Details
Gene Symbol: WARS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309323.1 | NP_001296252.1 | 4 - 5 | 525 | 69 | |
XM_023640270.1 | XP_023496038.1 | 556 | 69 | |||
GenBank mRNA | JL616080.1 | - | 4 - 5 | 529 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
