Assay Details
Gene Symbol: RSU1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309339.1 | NP_001296268.1 | 6 - 7 | 632 | 68 | |
XM_005606904.3 | XP_005606961.1 | 650 | 68 | |||
GenBank mRNA | JL616495.2 | - | 6 - 7 | 574 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
