Assay Details
Gene Symbol: PPP3R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309345.1 | NP_001296274.1 | 3 - 4 | 277 | 100 | |
XM_023618419.1 | XP_023474187.1 | 390 | 100 | |||
XM_023618420.1 | XP_023474188.1 | 426 | 100 | |||
GenBank mRNA | JL629851.1 | - | 1 - 2 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() |
