Assay Details
Gene Symbol: CCNG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309417.2 | NP_001296346.1 | 6 - 7 | 909 | 82 | |
GenBank mRNA | JL616062.2 | - | 6 - 7 | 912 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
