Assay Details
Gene Symbol: JKAMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309418.1 | NP_001296347.1 | 3 - 4 | 302 | 81 | |
XM_023627718.1 | XP_023483486.1 | 334 | 81 | |||
GenBank mRNA | JL616074.1 | - | 3 - 4 | 302 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
