Assay Details
Gene Symbol: FKBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309461.1 | NP_001296390.1 | 8 - 9 | 966 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FKBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309461.1 | NP_001296390.1 | 8 - 9 | 966 | 74 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |