Assay Details
Gene Symbol: RPS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309481.1 | NP_001296410.1 | 4 - 5 | 316 | 74 | |
GenBank mRNA | CX593239.1 | - | 4 - 5 | 339 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
