Assay Details
Gene Symbol: RPS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309481.1 | NP_001296410.1 | 5 - 6 | 419 | 63 | |
GenBank mRNA | CX593239.1 | - | 5 - 6 | 442 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
