Assay Details
Gene Symbol: KRT36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001346185.1 | NP_001333114.1 | 6 - 7 | 1253 | 80 | |
XM_023651628.1 | XP_023507396.1 | 2204 | 80 | |||
XM_023651629.1 | XP_023507397.1 | 2204 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
