Assay Details
Gene Symbol: CDC42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001309409.2 | NP_001296338.1 | 5 | 1822 | 100 | |
XM_023626612.1 | XP_023482380.1 | 1924 | 100 | |||
GenBank mRNA | JL615888.2 | - | 5 | 1773 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
