Assay Details
Gene Symbol: GABARAPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501541.5 | XP_001501591.1 | 399 | 72 | ||
GenBank mRNA | JL616126.1 | - | 3 - 4 | 356 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
