Assay Details
Gene Symbol: PTP4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503862.6 | XP_001503912.1 | 1047 | 71 | ||
XM_023634517.1 | XP_023490285.1 | 1306 | 71 | |||
GenBank mRNA | JL616021.2 | - | 4 - 5 | 985 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
