Assay Details
Gene Symbol: EIF3J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023618545.1 | XP_023474313.1 | 1417 | 82 | ||
XM_023618549.1 | XP_023474317.1 | 839 | 82 | |||
XM_023618559.1 | XP_023474327.1 | 665 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
