Assay Details
Gene Symbol: PLCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005600910.3 | XP_005600967.2 | 3230 | 61 | ||
XM_023620886.1 | XP_023476654.1 | 3318 | 61 | |||
XM_023620887.1 | XP_023476655.1 | 3208 | 61 | |||
XM_023620888.1 | XP_023476656.1 | 3313 | 61 | |||
XM_023620889.1 | XP_023476657.1 | 3328 | 61 | |||
GenBank mRNA | JL639207.1 | - | 1 - 2 | 807 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
