Assay Details
Gene Symbol: ZBTB16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014741177.2 | XP_014596663.1 | 2164 | 64 | ||
XM_014741178.2 | XP_014596664.1 | 2165 | 64 | |||
XM_023644848.1 | XP_023500616.1 | 2293 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
