Assay Details
Gene Symbol: HRAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023654737.1 | XP_023510505.1 | 474 | 68 | ||
XM_023654738.1 | XP_023510506.1 | 474 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
