Assay Details
Gene Symbol: RELT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023645768.1 | XP_023501536.1 | 1271 | 65 | ||
XM_023645769.1 | XP_023501537.1 | 1221 | 65 | |||
XM_023645770.1 | XP_023501538.1 | 10445 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
