Assay Details
Gene Symbol: ELK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023634178.1 | XP_023489946.1 | 520 | 59 | ||
XM_023634179.1 | XP_023489947.1 | 738 | 59 | |||
XM_023634180.1 | XP_023489948.1 | 648 | 59 | |||
XM_023634181.1 | XP_023489949.1 | 611 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
