Assay Details
Gene Symbol: CLNS1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494468.6 | XP_001494518.2 | 215 | 62 | ||
XM_023645653.1 | XP_023501421.1 | 217 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
