Assay Details
Gene Symbol: ARFIP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014741890.2 | XP_014597376.1 | 1057 | 80 | ||
XM_014741891.1 | XP_014597377.1 | 616 | 80 | |||
XM_023645903.1 | XP_023501671.1 | 883 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
