Assay Details
Gene Symbol: LOC106783453 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023645913.1 | XP_023501681.1 | 367 | 98 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023645913.1 transcripts.
Set Membership: |
![]() ![]() |
