Assay Details
Gene Symbol: LOC102147743 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023621809.1 | XP_023477577.1 | 323 | 87 | ||
XM_023621810.1 | XP_023477578.1 | 462 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023621809.1, XM_023621810.1 transcripts.
Set Membership: |
![]() ![]() |
