Assay Details
Gene Symbol: NIFK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023622708.1 | XP_023478476.1 | 309 | 79 | ||
XM_023622709.1 | XP_023478477.1 | 309 | 79 | |||
XM_023622710.1 | XP_023478478.1 | 309 | 79 | |||
GenBank mRNA | JL616615.1 | - | 2 - 3 | 261 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
