Assay Details
Gene Symbol: SNX24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005599478.3 | XP_005599535.1 | 931 | 69 | ||
XM_023617801.1 | XP_023473569.1 | 642 | 69 | |||
XM_023617802.1 | XP_023473570.1 | 414 | 69 | |||
XM_023617803.1 | XP_023473571.1 | 728 | 69 | |||
XM_023617804.1 | XP_023473572.1 | 577 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
