Assay Details
Gene Symbol: EFNA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023617907.1 | XP_023473675.1 | 1328 | 68 | ||
XM_023617908.1 | XP_023473676.1 | 1328 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
