Assay Details
Gene Symbol: POLR3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504618.4 | XP_001504668.1 | 781 | 100 | ||
XM_014730771.2 | XP_014586257.1 | 779 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
