Assay Details
Gene Symbol: PDE6D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003365136.4 | XP_003365184.2 | 333 | 94 | ||
XM_014740430.2 | XP_014595916.1 | 192 | 94 | |||
XM_023642558.1 | XP_023498326.1 | 459 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() |
