Assay Details
Gene Symbol: TENT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005599598.3 | XP_005599655.2 | 1094 | 63 | ||
XM_014730792.2 | XP_014586278.2 | 1094 | 63 | |||
XM_023618102.1 | XP_023473870.1 | 1096 | 63 | |||
XM_023618103.1 | XP_023473871.1 | 866 | 63 | |||
XM_023618104.1 | XP_023473872.1 | 1096 | 63 | |||
XM_023618106.1 | XP_023473874.1 | 1094 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
