Assay Details
Gene Symbol: NAIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504041.5 | XP_001504091.2 | 3558 | 117 | ||
XM_005599639.3 | XP_005599696.1 | 3556 | 117 | |||
XM_014730824.2 | XP_014586310.1 | 3556 | 117 |

More Information
Set Membership: |
![]() ![]() |
