Assay Details
Gene Symbol: NCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488049.6 | XP_001488099.1 | 585 | 62 | ||
XM_005599781.3 | XP_005599838.1 | 584 | 62 | |||
XM_023618487.1 | XP_023474255.1 | 630 | 62 | |||
XM_023618488.1 | XP_023474256.1 | 446 | 62 | |||
XM_023618489.1 | XP_023474257.1 | 447 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
