Assay Details
Gene Symbol: IL18R1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491301.5 | XP_001491351.3 | 907 | 72 | ||
XM_023618505.1 | XP_023474273.1 | 907 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
