Assay Details
Gene Symbol: SH2D6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023618797.1 | XP_023474565.1 | 1102 | 105 | ||
XM_023618798.1 | XP_023474566.1 | 1026 | 105 | |||
XM_023618799.1 | XP_023474567.1 | 1023 | 105 | |||
XM_023618800.1 | XP_023474568.1 | 988 | 105 | |||
XM_023618801.1 | XP_023474569.1 | 963 | 105 | |||
XM_023618802.1 | XP_023474570.1 | 1099 | 105 | |||
XM_023618803.1 | XP_023474571.1 | 1102 | 105 | |||
XM_023618804.1 | XP_023474572.1 | 1023 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
