Assay Details
Gene Symbol: MRPL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498597.6 | XP_001498647.2 | 258 | 83 | ||
XM_014731075.2 | XP_014586561.1 | 262 | 83 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MRPL19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498597.6 | XP_001498647.2 | 258 | 83 | ||
XM_014731075.2 | XP_014586561.1 | 262 | 83 |