Assay Details
Gene Symbol: HTRA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500046.5 | XP_001500096.1 | 1116 | 69 | ||
XM_014731077.2 | XP_014586563.1 | 1184 | 69 | |||
XM_023618835.1 | XP_023474603.1 | 1085 | 69 | |||
GenBank mRNA | JL618617.1 | - | 1 - 2 | 224 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
