Assay Details
Gene Symbol: ACTG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001487886.4 | XP_001487936.1 | 102 | 69 | ||
XM_023618878.1 | XP_023474646.1 | 103 | 69 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001487886.4, XM_023618878.1 transcripts.
Set Membership: |
![]() ![]() |
