Assay Details
Gene Symbol: DUSP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005599901.3 | XP_005599958.1 | 1002 | 66 | ||
XM_005599902.3 | XP_005599959.1 | 1001 | 66 | |||
XM_023618882.1 | XP_023474650.1 | 1007 | 66 | |||
XM_023618883.1 | XP_023474651.1 | 2388 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
