Assay Details
Gene Symbol: PELI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493946.6 | XP_001493996.1 | 1331 | 62 | ||
XM_023619004.1 | XP_023474772.1 | 1105 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
