Assay Details
Gene Symbol: EFEMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001917597.4 | XP_001917632.2 | 692 | 59 | ||
XM_005599971.3 | XP_005600028.2 | 657 | 59 | |||
XM_023619660.1 | XP_023475428.1 | 741 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
