Assay Details
Gene Symbol: GALM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500603.4 | XP_001500653.1 | 589 | 71 | ||
XM_023619209.1 | XP_023474977.1 | 590 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
