Assay Details
Gene Symbol: KIF3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502843.4 | XP_001502893.1 | 1793 | 72 | ||
XM_005600158.3 | XP_005600215.1 | 1793 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
