Assay Details
Gene Symbol: SF3B6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503183.3 | XP_001503233.1 | 408 | 76 | ||
GenBank mRNA | JL615853.2 | - | 2 - 3 | 163 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
