Assay Details
Gene Symbol: LDAH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001501695.5 | XP_001501745.3 | 933 | 79 | ||
XM_014731290.2 | XP_014586776.2 | 852 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
