Assay Details
Gene Symbol: COLEC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001502880.5 | XP_001502930.2 | 1117 | 51 | ||
XM_014731318.2 | XP_014586804.1 | 1117 | 51 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
