Assay Details
Gene Symbol: ISY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001488818.4 | XP_001488868.1 | 520 | 73 | ||
GenBank mRNA | JL619623.1 | - | 1 - 2 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
