Assay Details
Gene Symbol: MRPS25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491163.4 | XP_001491213.3 | 439 | 100 | ||
XM_023619923.1 | XP_023475691.1 | 706 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
