Assay Details
Gene Symbol: MBD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014731405.2 | XP_014586891.1 | 300 | 68 | ||
XM_014731406.2 | XP_014586892.1 | 301 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
