Assay Details
Gene Symbol: CIDEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005600382.3 | XP_005600439.1 | 158 | 78 | ||
XM_023619999.1 | XP_023475767.1 | 926 | 78 | |||
XM_023620000.1 | XP_023475768.1 | 925 | 78 | |||
XM_023620001.1 | XP_023475769.1 | 255 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
