Assay Details
Gene Symbol: RYBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023620086.1 | XP_023475854.1 | 260 | 82 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
