Assay Details
Gene Symbol: ABHD14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494476.3 | XP_001494526.1 | 320 | 97 | ||
XM_014731640.2 | XP_014587126.1 | 335 | 97 | |||
XM_023620352.1 | XP_023476120.1 | 1142 | 97 | |||
XM_023620353.1 | XP_023476121.1 | 433 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
