Assay Details
Gene Symbol: CISH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001493420.5 | XP_001493470.2 | 388 | 66 | ||
XM_005600615.3 | XP_005600672.1 | 439 | 66 | |||
XM_005600616.3 | XP_005600673.1 | 1405 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
